Glass microfibre filters for personal air samplers, grade GF/A, Whatman™
Fournisseur: Whatman products (Cytiva)
18208296
513-0105EA 155 EUR InStock
513-0105
Glass microfibre filters for personal air samplers, grade GF/A, Whatman™
Filtres Filtres de fibres de verre
Grade GF/A card-mounted filters are used in static sample and personal air sampler applications. These aerosol sampling and particulate monitoring filters provide high flow rates and minimal sample interference.
  • Heat resistant up to 500 °C
  • Made of high quality borosilicate glass fibres without added binders
  • Low moisture affinity and high loading capacity
  • Card mounted for use with a continuous air monitor or personal air sampler
Accurate suspended particulate matter analysis is critical in nuclear plants and similar environments.
The GF/A card mounted filter has dimensions of 55×37,5 mm. Please verify that your personal air sampler supports a sample card of this size.
Certifications: Manufactured according to ISO9001:2008 QMS standards.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité