Vacuum filter holder, Sartolab® Multistation
Fournisseur: Sartorius
SDLC01
514-1196EA 3050 EUR InStock
514-1196
Vacuum filter holder, Sartolab® Multistation
Porte-filtres
Sartolab® MultiStation is a stand especially designed to hold one to six vacuum filtration units. It allows the simultaneous filtration of multiple samples.
  • Hands-free filtration
  • Time saving (no extra installation time to vacuum for each single filter unit)
The MultiStation is permanently connected to a vacuum source; the vacuum filtration units can be easily installed in the MultiStation for quick and easy filtration of samples without extra time consuming
connection.
Sartolab® MultiStation functions with the Sartolab® RF 50 vacuum filtration unit. The adapter connecting the funnel to the 50 ml sample tube fits perfectly into the MultiStation bracket.
One click and the filtration unit hangs in the device. That ensures perfect stability of the filter; with a second click, the vacuum is automatically applied and filtration begins.
The rotating head is multi-directional, enabling you to easily handle your samples in front of you. Furthermore, all six brackets are numbered to avoid any confusion during the filtration of multiple samples.
Commander maintenant
COMMANDE
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité