Grade 2589 C filter papers for technical use, Whatman™
Fournisseur: Whatman products (Cytiva)
10343876
515-8080EA 68.4 EUR InStock
515-8080
Grade 2589 C filter papers for technical use, Whatman™
Filtres Papiers filtre Papiers filtres qualitatifs
Thick filter with medium to slow filtration speed, high wet strength, and good retention for smaller particles.
These filter papers are made from super-refined cellulose and have been specifically designed to have particular properties for each technical application. The applications range from the filtration of beverages to the purification of electroplating baths.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité