Acro® 500 Filter Capsule with Supor® EKV Membrane and Polycarbonate Housing
Fournisseur: Cytiva (Formerly Pall Lab)
12337
516-1169EA 0 EUR InStock
516-1169
Acro® 500 Filter Capsule with Supor® EKV Membrane and Polycarbonate Housing
Filtres Capsules de filtration
The Acro 500 filter capsule with Supor EKV membrane and polycarbonate housing offers a 500 cm² effective filtration area (EFA) for the sterile filtration of larger volumes of laboratory solutions.
  • Membrane features a built-in dual layer polyethersulfone (PES) construction which provides fast and efficient filtration
The low protein and preservative binding properties of the membrane allow for maximum transmission of active ingredients as well as broad liquid compatibility, ideal for the filtration of a wide range of fluids including buffers,More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité