51779-1L
51779-1LEA
165
EUR
InStock
51779-1L
Diméthylsulfoxyde ≥99.9% (par GC), CHROMASOLV™ en phase gazeuse Headspace, Riedel-de Haën™
Diméthylsulfoxyde
Honeywell Chromasolv™ GC-headspace solvents:
- Developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
- Very pure solvents with extremely low concentrations of the defined residual solvents