Lewatit® AF 5, Pore size: 20 Å (mean), 20-50 mesh, Supelco®
Fournisseur: Merck
Synonyms: Lewasorb AF 5
52089-500G-F
52089-500G-FEA 469 EUR InStock
52089-500G-F
Lewatit® AF 5, Pore size: 20 Å (mean), 20-50 mesh, Supelco®
Lewatit® AF 5
Lewatit® AF 5 is a microporous carbonaceous bead type material designed for downstream process separation and purification. It is derived from a synthetic polymer.
  • Surface area: 1300 m²/g
Lewatit® AF 5 is used for adsorptive polishing in water treatment applications, and for adsorption for traces of organic substances such as chlorinated hydrocarbons, MTBE, organic phosphates, amines, pesticides, herbicides, and metabolites. Lewatit® AF 5 is remarkable for its well defined uniform pore distribution, high surface area, and high adsorptive capacity. The separation mechanism is partially via low energy hydrogen bonding, resulting in simple elution methods with a variety of solvents. Lewatit® AF 5 is eluted with steam, or hot water.
Lewatit® AF may be used as separation media for HPLC, UPLC and paper chromatography.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité