Rotors for Centrifuge 5425/5425 R
Fournisseur: EPPENDORF
5495500.006 5495503.005 5495505.008 5495508.007 5495510.001 5495512.004
521-2481EA 742 EUR InStock
521-2481 521-2482 521-2483 521-2484 521-2485 521-2486
Rotors for Centrifuge 5425/5425 R
Centrifugeuses Rotors
Eppendorf rotors for centrifuge 5425/5425 R, precision meets versatility. Optimise your centrifuge with a range of capacities and speeds. Explore accessory options such as adaptors and buckets for tailored sample processing, ensuring optimal results in molecular biology and beyond.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 minutes)
  • Accessories (adaptors and buckets) further customise your centrifuge setup
  • Aerosol-tight and PTFE coated options available
Refine your centrifugation experience with Eppendorf rotors designed for centrifuge 5425/5425 R. Offering versatility in capacities and speeds, these rotors cater to a spectrum of applications, from microcentrifugation to larger sample volumes. Accessories such as adaptors and buckets further customise your setup for specific needs. Achieve reliable sample separation with precision and flexibility, meeting the demands of molecular biology, genomics, and beyond. Explore the comprehensive range to optimise your centrifuge for reproducible results.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité