Rotors for Centrifuges, 5920 R
Fournisseur: EPPENDORF
5895101.003 5895117.007 5895118.003 5895100.007 5895190.006
521-2490EA 1446 EUR InStock
521-2490 521-2498 521-2499 521-2489 521-2509
Rotors for Centrifuges, 5920 R
Centrifugeuses Rotors
Eppendorf Rotors for Centrifuge 5920 R - precision in refrigerated large-capacity centrifugation. Tailor your experiments with varied capacities and speeds. Accessorize with versatile buckets and adaptors for efficient sample processing. Ideal for high-throughput applications, ensuring accuracy and reproducibility.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
  • Aerosol-tight options available
Experience precision in refrigerated large-capacity centrifugation with Eppendorf rotors designed for centrifuges 5920 R. These rotors offer a range of capacities and speeds, providing flexibility for various sample processing needs. Accessorise with versatile buckets and adaptors, enhancing the adaptability of your centrifuge setup. Ideal for high-throughput applications, the centrifuges 5920 R is further optimized with these precision-engineered rotors, ensuring accuracy and reproducibility in every spin. Explore the range to elevate your refrigerated large-capacity centrifugation experience.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
SERVICES
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : technical.service.be@vwr.com
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité