Microcentrifuge, Refrigerated, 5427 R (IVD marked)
Fournisseur: EPPENDORF
5429000.015
521-4270EA 6555.95 EUR InStock
521-4270
Microcentrifuge, Refrigerated, 5427 R (IVD marked)
Centrifugeuses Microcentrifugeuses
This fast running microcentrifuge with new natural refrigerant (R 290, propane) and significantly lower Global Warming Potential is designed for maximum application flexibility and offers a versatile choice of rotors in order to serve different user demands.
  • End of run timer, brightly lit display and more precise adjustment of time in the 1st minute (in 5 second steps)
  • One-Push-Short Spin button for quick centrifugation
  • Max. RCF: 25001 xg (16220 rpm)
  • FastTemp function for rapid pre-cooling of the centrifuge, e.g. from 23 to 4 °C in approx. 11 minutes
  • ECO-shut off of the compressor for a lower energy consumption when not in use
  • QuickLock® rotor lid for all aerosol-tight rotors
There are 9 different rotors available, including high capacity rotors with up to 48 places as well as high speed rotors for a quick sample throughput
Informations de commande: Can be supplied with or without rotor.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
SERVICES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité