Synonyms:
1,1,1,3,3,3-Hexafluoroisopropyl alcohol, Hexafluoro-2-propanol, Hexafluoroisopropanol, 1,1,1,3,3,3-Hexafluoropropan-2-ol, HFIP
52517-10ML
52517-50ML
52517-10X1ML
52517-10MLEA
222
EUR
InStock
52517-10ML
52517-50ML
52517-10X1ML
1,1,1,3,3,3-Hexafluoro-2-propanol ≥99.8%, LiChropur™ pour dérivatisation GC, Supelco®
1,1,1,3,3,3-Hexafluoro-2-propanol
Hexafluoroisopropanol (HFP) is generally used to denature the native state of proteins, and it also stabilizes the a-helical conformation in unfolded peptides and proteins like ß-lactoglobulin and melittin
Suitable for the derivatization of glutamic acid, homovanillic, iso-homovanillic and dihydroxyphenyl acetic acids and Υ-aminobutyric acid.