SPE tubes, Supelclean™ Florisil®/Na₂SO₄
Fournisseur: Merck
52582-U
52582-UEA 669 EUR InStock
52582-U
SPE tubes, Supelclean™ Florisil®/Na₂SO₄
Colonnes pour chromatographie
Dual layer SPE tube that contains Na2SO4 (upper layer) and Florisil (magnesium silicate; lower layer) separated and packed with PTFE frits (glass tubes) or PE frits (PP tubes).
  • Resistant to harsh chemicals and aggressive solvents
  • Absence of leachables such as pthalates and plasticizers
  • Florisil particle size- 60/100 mesh (150 to 200 μm); Na2SO4 Purity - 99,99+%, density - 2,68 g/ml
  • Excellent for removing/isolating polar compounds from organic matrices
  • Na₂SO₄ layer aids in removing aqueous sample residues that may hinder Florisil performance and/or subsequent GC analysis
Available in glass SPE hardware allowing users to reactivate Florisil through heating at 140 °C, 16 hours.
Use in conjunction with Visiprep Large Volume Sampler and Visiprep SPE Vacuum Manifolds for processing larger volume samples.
Conditionnement: Pack of 48.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité