Diaphragm vacuum pump, VACUUBRAND®
Fournisseur: LABCONCO
7393001
531-0942EA 6260 EUR InStock
531-0942
Diaphragm vacuum pump, VACUUBRAND®
Pompe Pompes à vide à membrane
Single- or dual-head chemically resistant, dry running diaphragm pumps for a wide range of laboratory applications. They transfer and evacuate without affecting the media, i.e. media remains uncontaminated.
  • 100% oil-free displacement, so media cannot be contaminated by transfer, evacuation or compression
  • High resistance to vapours and condensates
  • Chemically resistant, suitable for use with very aggressive and corrosive gases and vapours
  • Maintenance-free, environmentally friendly; the pumps use no water and so produce no contaminated waste water
Application: Fluid aspiration, degassing, gel drying, rotary evaporation, centrifugal concentration, distillation
Includes diaphragm with patented tension optimisation, which allows the pump size to be smaller and extends the service life of the diaphragm. All parts that come into contact with the media are comprised of chemically resistant fluorinated plastics.
Informations de commande: Includes chemically resistant diaphragm pump with thermal circuit breaker, mains fuse, connecting cable and PTFE hose attachments with for hoses with 10 mm internal diameter.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
SERVICES
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : technical.service.be@vwr.com
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité