Torque-measuring stirrer, STARVISC 200-2.5 control
Fournisseur: IKA
25003604
531-1108EA 7960 EUR InStock
531-1108
Torque-measuring stirrer, STARVISC 200-2.5 control
Agitateurs Agitateurs à tige
The STARVISC 200-2.5 control stirrer measures viscosity and displays it. The result can be read in real-time on the display. It has a broad range of applications.
  • High-precision and trend torque measurement
  • A viscosity calculation can be carried out immediately via a user friendly menu
  • Removable control unit and TFT display
  • Stepless speed control
  • Even highly viscous substances can be intensively stirred using the powerful STARVISC stirrer
This is particularly helpful during product development, clearly indicates while running research programmes as to whether the stirred substance can be used as desired.
Certifications: IP protection class according to DIN EN 60529: IP 40
Informations de commande: Different stirrers tools can be ordered seperately.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité