Solid phase extraction cartridges, Bond Elut™ Plexa
Fournisseur: VARIAN
12109206 12109303 12109301 12109601 12109603 12259506 327832 12109301T 12109703 12169610B 12281305
549-0811EA 197 EUR InStock
549-0811 549-0814 549-0812 549-0815 549-0816 549-0839 VARI327832 549-0813 549-0817 549-0819 VARI12281305
Solid phase extraction cartridges, Bond Elut™ Plexa
Colonnes pour chromatographie
The water-wettable, hydroxylated exterior allows excellent flow, even with biological fluids. A gradient of polarity on the polymer surface shunts small analytes to the more hydrophobic centre of the polymer bead, where they are retained prior to the washing and elution steps. Plexa provides these performance enhancements due to a unique polymeric architecture with a non-retentive, hydroxylated, amide-free surface and a non-polar PS/DVB core for retaining small molecules.
  • Fast flow, reproducible performance
  • Improved extract cleanliness minimises sample matrix interferences
  • Non-polar retention mechanism
Binding of proteins and lipids on the polymer surface is minimised, resulting in cleaner samples and reduced matrix interference. Plexa is ideal for high-throughput tests requiring validated performance with minimal method development. The standard non-polar retention mechanism is applicable to almost any analyte type. The performance features operate at the sample loading step, making them largely method independent.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité