Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Acide sulfurique 95.0-98.0% (par acidimétrie), AR Select® ACS, Macron Fine Chemicals™

Fournisseur: Avantor Performance Materials
Danger

Management of Change (MOC) category = N

5557-14EA 96.37 EUR InStock
5557-14
Acide sulfurique 95.0-98.0% (par acidimétrie), AR Select® ACS, Macron Fine Chemicals™
Acide sulfurique


Formule: H₂SO₄
Point d'ébullition: ∼335 °C (1013 hPa)
Point de fusion: 3 °C
Densité: 1.84 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00064589
Numéro CAS: 7664-93-9
EINECS: 231-639-5
ONU: 1830
ADR: 8,II
Merck Index: 14,08974
Commander maintenant
Résultats des tests de spécification
Meets ACS Reagent Chemical Requirements 95.0 - 98.0%
For Laboratory,Research,or Manufacturing Use ≤0.5 ppm
ACS - Assay (H₂SO₄) (by acidimetry) 95.0 - 98.0 %
Appearance Passes Test
ACS - Ammonium (NH₄) ≤ 0.5 ppm
ACS - Arsenic and Antimony (as As) ≤ 0.003 ppm
ACS - Chloride (Cl) ≤ 0.05 ppm
ACS - Color (APHA) ≤ 8
ACS - Heavy Metals (as Pb) ≤ 0.1 ppm
ACS - Nitrate (NO₃) ≤ 0.1 ppm
ACS - Residue after Ignition ≤ 1 ppm
ACS - Substances Reducing Permanganate (as SO₂) ≤ 2 ppm
Determine by Flame Photometry & ICP - Aluminum (Al) ≤ 50.0 ppb
Determine by Flame Photometry & ICP - Barium (Ba) ≤ 10.0 ppb
Determine by Flame Photometry & ICP - Beryllium (Be) ≤ 50.0 ppb
Determine by Flame Photometry & ICP - Bismuth (Bi) ≤ 100.0 ppb
Determine by Flame Photometry & ICP - Boron (B) ≤ 10.0 ppb
Determine by Flame Photometry & ICP - Cadmium (Cd) ≤ 5.0 ppb
Determine by Flame Photometry & ICP - Calcium (Ca) ≤ 50.0 ppb
Determine by Flame Photometry & ICP - Chromium (Cr) ≤ 10.0 ppb
Determine by Flame Photometry & ICP - Cobalt (Co) ≤ 5.0 ppb
Determine by Flame Photometry & ICP - Copper (Cu) ≤ 5.0 ppb
Determine by Flame Photometry & ICP - Gallium (Ga) ≤ 100.0 ppb
Determine by Flame Photometry & ICP - Germanium (Ge) ≤ 100.0 ppb
Determine by Flame Photometry & ICP - Gold (Au) ≤ 100.0 ppb
Determine by Flame Photometry & ICP - Iron (Fe) ≤ 50.0 ppb
Determine by Flame Photometry & ICP - Lead (Pb) ≤ 1.0 ppb
Determine by Flame Photometry & ICP - Lithium (Li) ≤ 10.0 ppb
Determine by Flame Photometry & ICP - Magnesium (Mg) ≤ 50.0 ppb
Determine by Flame Photometry & ICP - Manganese (Mn) ≤ 5.0 ppb
Determine by Flame Photometry & ICP - Mercury (Hg) ≤ 1.0 ppb
Determine by Flame Photometry & ICP - Molybdenum (Mo) ≤ 10 ppb
Determine by Flame Photometry & ICP - Nickel (Ni) ≤ 5.0 ppb
Determine by Flame Photometry & ICP - Potassium (K) ≤ 50.0 ppb
Determine by Flame Photometry & ICP - Silicon (Si) ≤ 100.0 ppb
Determine by Flame Photometry & ICP - Silver (Ag) ≤ 10.0 ppb
Determine by Flame Photometry & ICP - Sodium (Na) ≤ 500.0 ppb
Determine by Flame Photometry & ICP - Strontium (Sr) ≤ 10.0 ppb
Determine by Flame Photometry & ICP - Thallium (Tl) ≤ 200.0 ppb
Determine by Flame Photometry & ICP - Tin (Sn) ≤ 50.0 ppb
Determine by Flame Photometry & ICP - Vanadium (V) ≤ 10 ppb
Determine by Flame Photometry & ICP - Zinc (Zn) ≤ 5.0 ppb
Determine by Flame Photometry & ICP - Zirconium (Zr) ≤ 10 ppb
Determine by Flame Photometry & ICP

 

COMMANDE

Documentation

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité