Organic reference standard, Acetaldehyde diethyl acetal, analytical standard, Supelco®
Fournisseur: Merck
55795-1ML
55795-1MLEA 65.5 EUR InStock
55795-1ML
Organic reference standard, Acetaldehyde diethyl acetal, analytical standard, Supelco®
Étalons Étalons organiques
Acetaldehyde diethyl acetal is one of the precursors utilised for the synthesis of fragrances and pharmaceutical products.
Acetaldehyde diethyl acetal may be used as a reference standard for the determination of acetaldehyde diethyl acetal in: wine vinegars by headspace sorptive extraction−thermal desorption−gas chromatography−mass spectrometry (HSSE−TD−GC−MS); distilled spirits by GC with flame ionisation detection (FID) as well as GC-FID-MS; organic red wines derived from Saccharomyces cerevisiae strains by GC-FID as well as HSSE-GC−MS; cognac (brandy) by HS-GC-MS.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fd0d5cc73c15775087e45ab44f2aeb7976adddf&vid=b1c476b2eeabe59ca3a026dc6d464f2d5e051781&prid=web&clid=SAPDAM
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité