Management of Change (MOC) category = NI
5611-02EA
70.15
EUR
InStock
5611-02
Acide chlorhydrique 0.01 N, BAKER ANALYZED® solution volumétrique, J.T.Baker®
Acide chlorhydrique
Standardized at 25 °C against chemicals whose certification is traceable to NIST. Lot standardization value accurate to ±1 part per 1000, provided on label.