Management of Change (MOC) category = NI
5616-02EA
78.77
EUR
InStock
5616-02
Acide chlorhydrique 2 N, BAKER ANALYZED® solution volumétrique, J.T.Baker®
Acide chlorhydrique
Standardised at 25 °C against chemicals whose certification is traceable to NIST. Lot standardisation value, accurate to ±1 part per 1000, provided on label.