Air compressor, Solaris
Fournisseur: PEAK SCIENTIFIC
65-0003
564-0278EA 5460 EUR InStock
564-0278
Air compressor, Solaris
Générateurs de gaz Stations d'air comprimé
This air compressor generator has been designed to support the Solaris nitrogen generator for ELSD in laboratories without an external supply of high quality air.
  • Stackable with Solaris nitrogen generator for ELSD and Compact Mass Specs
  • Designed specifically for ELSD and Compact Mass Specs
Solaris has been developed to provide an on-site gas supply of high purity nitrogen for ELSD for laboratories using HPLC or alternatively to supply Compact Mass Spectrometer instruments.
Min./max. operating temperature: 5 to 35 °C
Max. pressure: 8,27 bar
Gas outlet: 1×¼" BSPP
Certifications: Manufactured and tested to highest spec in UK, CE/FCC certified.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité