Portable Label Printer Kit EU and Brady Workstation Suite, M410
Fournisseur: Brady
322718
568-0930EA 585 EUR InStock
568-0930
Portable Label Printer Kit EU and Brady Workstation Suite, M410
Imprimantes Label Printers
Specifically engineered for premium 25,40 mm printing, the M410 label printer is an everyday workhorse for DataCom, electrical and lab professionals.
  • Print pre-sized or continuous labels up to 25,40 mm wide with edge-to-edge printing
  • 110+ hassle-free drop-in cartridges with 12+ purpose-built materials to take on whatever your work demands
  • Get seamless cartridge changes with an easy-swap locking mechanism while a window in the door allows you to easily remember what's installed
  • Create and print two ways - the intuitive QWERTY keypad or a PC
  • Designed to withstand 1,2 m drops and challenging work environments
  • Enhanced exterior colouring for jobsite visibility
  • Ships with a licence for the Brady workstation suite of your preference: Product and wire identification, safety and facility identification, or lab identification, enabling easy creation of a wide range of labels and signs
  • Backlit LCD graphics display for easier labelling in low-lit areas
Applications include: Asset tracking, component and equipment labelling, data and telecommunications labelling, electrical labelling, facility identification, general identification, inventory and inspection labelling, laboratory labelling, wire and cable labelling.
Cutter type: Single hand manual actuate, straight cut, with label retention.
Certifications: UKCA.
Informations de commande: Two-year warranty. This M410 printer ships in a hardcase with a rechargeable NiMH battery pack, an AC adapter, B-595 vinyl label cartridge, a magnet, a utility hook, a quick start guide, a USB cable, a licence for Brady workstation software suites and a USB flash drive with driver and user manual.
Conditionnement: Hard case.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité