Thapsigargin ≥97% (par CCM), solide incolore, Millipore®
Fournisseur: Merck Millipore Calbiochem
586005-1
586005-1EA 262 EUR InStock
586005-1
Thapsigargin ≥97% (par CCM), solide incolore, Millipore®
Thapsigargin
Thapsigargin is a cell-permeable tumour-promoting sesquiterpene lactone that releases Ca²⁺ by inhibiting endoplasmic reticular Ca²⁺-ATPase.
  • Colourless solid
It does not increase inositol phosphates and has no significant effect on protein kinase C. It increases Ca²⁺-dependent Na⁺ influx in human platelets in a dose-dependent manner. Ii induces apoptosis in rat thymocytes and in human hepatoma cells and in alveolar rhabdomyosarcoma cells by elevating Akt-stimulated PAX3-FOXO1 phosphorylation and promoting its proteasomal degradation.
Formule: C₃₄H₅₀O₁₂
Température de stockage: Congélateur
Numéro MDL: MFCD00083511
Numéro CAS: 67526-95-8
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité