VWR®, Micro Anti-Static Weighing Boats
Fournisseur: VWR Collection
611-5050EA 38.2 EUR InStock
611-5050
VWR®, Micro Anti-Static Weighing Boats
Nacelles Nacelles de pesée
Ideal for microbalance weighing and/or distribution of very small amounts of sampling material.  
  • Large tabs make boats easy to use, position and hold, especially with forceps
  • Inert material enables the boat and contents to be added to solutions to aid 100% sample recovery, where protocols allow
  • Anti-static properties prevent sample retention from static electricity
Ideal for use with pipette calibration. Smaller surface area reduces evaporation rate.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité