Cubis® II Balances
Fournisseur: Sartorius Balances
MCA125S-U-MTS
611-5824EA 14010 EUR InStock
611-5824
Cubis® II Balances
Balances
The modular weighing system Cubis® II can be individually configured from different components. The combinability of the display unit, weighing module, draft shield, software packages for various applications and functions and a comprehensive range of accessories allows the individual adaptation of the Cubis® II balance to all weighing tasks.
  • Leading performance
  • Monolithical weighing system, integrated climate sensors, individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-levelling
  • Full end-to-end data integrity
  • Integrated audit-trail, state-of-the-art user management
  • Outstanding service support
  • Integrated status centre, integrated service functions preventive maintenance on accredited standards
The Cubis® II range of premium laboratory balances with a maximum load between 2,1 g and 70 kg and a readability between 0,1 μg to 1 g provide the ideal model for every application.
Connection type: D-sub connector, 9-pin
Display type: Colour touchscreen
Certifications: 21 CFR Part 11 compliance.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité