Moisture Analyser, MB92
Fournisseur: OHAUS
30971924
611-5850EA 3500 EUR InStock
611-5850
Moisture Analyser, MB92
Analyseurs Analyseurs d'humidité
High performance moisture analysis with advanced features for the most demanding application.
  • Robust and durable with the new carbon heating technology and an upgraded full colour capacitive touchscreen
  • MB92 is the perfect choice for users demanding a high level of performance and precision
  • Multiple drying profiles, shut off criteria and methods offer great flexibility in method creation
  • Enhanced password protection ensures greater data security and integrity
  • A full glass panel and tool-free heating chamber assembly makes cleaning and maintenance easy
  • Perfect for both the laboratory and the production floor
  • External calibration
The new MB92 features a redesigned housing, innovative carbon fiber heating and upgraded load cell for enhanced durability and performance. Ideal for users demanding a higher level of performance on both the laboratory and production floor, offering greater data security and integrity with password protection.
Certifications: IEC/EN 61010-1; IEC/EN 61010-2-010; IEC/EN 61326-1 Class B, Basic environments; CSA; RCM; WEEE.
Information de livraison: Includes pan handle, in-use cover, sample pans and glass fibre pads. Supplied with plastic in use protective cover.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité