Pipette controller, VITLAB maneus®
Fournisseur: VITLAB
1630500
612-4181EA 81.3 EUR InStock
612-4181
Pipette controller, VITLAB maneus®
Pipettes Pipeteurs
The VITLAB maneus® pipette controller enables both left- and right-handers to operate all normal bulb and graduated pipettes easily and fatigue-free. For use with all normal bulb and graduated pipettes from 0,1 to 200 ml.
  • Safe and easy handling allows even inexperienced users to adjust the meniscus precisely
  • Easy and fast replacement of the hydrophobic membrane filter
  • Optimised valve system: Liquids can be drawn up simply, without exerting pressure
  • Suction element provides rapid filling of the pipette (capacity: 50 ml in less than 10 seconds)
  • Discharge bellows for the emptying (blow-out) of the pipette
  • Specially moulded intake cone ensures secure seating for all normal bulb and graduated pipettes
  • Easy to dismantle, easy to clean, and completely autoclavable at 121 °C (2 bar)
Information de livraison: Supplied with replacement 3 µm membrane filter and instruction manual.
Commander maintenant
COMMANDE
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité