Reagent reservoirs for multi channel pipettes
Fournisseur: BDP International Inc.
4304 4305 4312 4322 4332
613-3186EA 36.9 EUR InStock
613-3186 613-3187 613-3190 613-3192 613-3194
Reagent reservoirs for multi channel pipettes
Réservoirs pour réactifs
Clear PS, sterile. These reservoirs have been designed to nest inside each other, making it possible to get twice as many reservoirs in half the space of other products on the market, reducing inventory space requirements and shipping costs. The reservoirs are made of crystal clear PS and fit into a reusable, white base with bold graduation markings.
  • Clearly visible graduations become invisible below fluids, making it easier to identify the exact desired volume line
  • Reusable base holds disposable reservoir securely during use; one free base is included with every case of bulk or individually sealed reservoirs
  • Base accommodates two reservoirs; one acting as a lid to allow short term storage of reagent
  • Pour back spouts molded into each reagent reservoir
  • Full length, extra deep trough is easily accessible for pipette tips: For maximum fluid recovery and minimal waste
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité