VWR®, Pipettes monocanales, électroniques, volume variable
Fournisseur: VWR Collection
613-5415EA 560 EUR InStock
613-5415 613-5416 613-5417 613-5418 613-5781
VWR®, Pipettes monocanales, électroniques, volume variable
Pipettes Micropipettes
These pipettes combine superior accuracy, precision and robustness with advanced ergonomic design.
  • Ease of use: Only two buttons and five pipetting modes; enables an easy one-handed programming and a quick pipetting cycle start
  • CUSTOM mode: Users can quickly and easily create specific protocols thanks to VWR® Protocol Builder software
  • Advanced ergonomics: Lightweight and perfectly balanced, it requires virtually no force to significantly reduce RSI
  • Fits comfortably in either the left or right hand and allows a 360° rotating manifold
  • Fully motorised piston drive: For even and regular sample delivery across each channel
  • Tip holders are designed for a secure tip fit and easy ejection thanks to a curved ejector bar
  • Pipettes can be used while charging without compromising performance
VWR® electronic pipettes offer outstanding performance in all pipetting modes. They require virtually no force to aspirate and dispense samples, and significantly reduce RSI (Repetitive Strain Injury) associated with routine work.
Modes de pipetage : PIPET (mode standard), REVERSE, REPETITIVE, MIX et CUSTOM.
Informations de commande: Each pipette is supplied with with one Lithium-ion battery, USB power supply (5 V), tip ejector extension (for 10 µl models only), lubricant (except for 10 µl models), certificate of conformity and a quick start guide. Accessory stands are sold separately.
Commander maintenant
COMMANDE
ACCESSOIRES
SERVICES
Services: Pour un étalonnage accrédité selon ISO8655 et ISO17025, un entretien ou une réparation, contactez-nous : technical.service.be@vwr.com
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité