ep Dualfilter T.I.P.S.® SealMax® Racks Self-Sealing Pipette Tips
Fournisseur: EPPENDORF
0030078.691 0030078.705 0030078.713 0030078.721 0030078.730 0030078.748
613-6791EA 207 EUR InStock
613-6791 613-6792 613-6793 613-6794 613-6795 613-6796
ep Dualfilter T.I.P.S.® SealMax® Racks Self-Sealing Pipette Tips
Pointes de pipettes
Eppendorf ep Dualfilter T.I.P.S.® SealMax® Racks provide advanced self-sealing pipette tips in 'PCR clean and Sterile' grade, featuring unique two-phase filters for unparalleled protection against contamination in sensitive lab applications.
  • Reliable protection from liquids, aerosols, and biomolecules
  • No PCR inhibiting additives
  • Protection of pipette from aggressive liquids when over-aspiration occurs
  • Available in PCR clean/Sterile (sterile and pyrogen-free)
  • Continuous inspection of each production lot by an independent laboratory guarantees high purity and quality
  • Modern slim rack design with reclosable lid
  • Improved sustainability through 20% lower polypropylene consumption per rack
  • ACT® labelled by non-profit My Green Lab® to help make an informed choice regarding sustainability
Superior protection for both sample and pipettes. The ep Dualfilter T.I.P.S.® SealMax® self-sealing filter pipette tips offer almost 100 % protection from contaminating aerosols and biomolecules with a two-phase design that works together to ensure optimal pipetting. Over-pipetted samples are stopped in their tracks with the instant closure of the Dualfilter. The white layer retains drops and splashes while the violet layer forms a barrier.
ep Dualfilter T.I.P.S.® SealMax® are ideally suited for methods where high sensitivity and reproducibility are essential. Both ep Dualfilter T.I.P.S.® and ep Dualfilter T.I.P.S.® SealMax® are especially recommended for applications such as PCR and microbiology, where preserving sample integrity and preventing cross contamination are critical.
The ep Dualfilter T.I.P.S.® SealMax® self-sealing filter pipette tips are manufactured to the highest quality under cleanroom conditions, without the use of interfering slip agents, biocides, and plasticisers. Made from the purest, non-recycled materials, self-sealing filter pipette tips are ideal for molecular and microbiological applications that require the greatest sensitivity and reproducibility, without the fear of sample degradation and cross-contamination.
Certifications: Lot-specific certficates for the purity grade PCR clean/Sterile can be downloaded from Eppendorf website. Certificate of filter efficiency of class ISO 25 E in accordance with DIN EN ISO 29463-5 available.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité