VWR®, Digital Burette
Fournisseur: VWR Collection
613-7002EA 650 EUR InStock
613-7002
VWR®, Digital Burette
Burettes
State-of-the art motor-operated burette with three calibrated pre-set speeds and accuracy with precision conforming to ISO 8655 standards.
  • Motor-controlled piston movement
  • Can be flexibly operated at three calibrated pre-set speeds: Fast, medium and dropwise
  • Recirculation mode to prevent the loss of reagent during purging
  • Flexible delivery nozzle to change angle of dispensing
  • User-friendly TFT touch screen
  • Handheld stylus provided as an accessory to assist touch operations
  • Powered by rechargeable, replaceable long lasting Li-ion battery
  • Up to 20 results can be stored in the instrument memory
  • Computer connectivity through a micro USB cable
The VWR® E-BURETTE is a motor-operated burette with sophisticated features and functionality. Motor-controlled piston movement eliminates errors associated with manual wheel controlled piston movement, thus facilitating correct and comfortable titrations. Having the capacity of 50 ml, this burette can be flexibly operated at three calibrated pre-set speeds, including dropwise dispensing for fine volume adjustment. This helps in achieving accurate end point of the titration experiment.
Commander maintenant
COMMANDE
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité