Filter tips
Fournisseur: BIOTIX
BT10 BT20 63300759 BT1250 BT1250.N
614-1419EA 480 EUR InStock
614-1419 614-1422 614-1424 BITXBT1250 BITXBT1250.N
Filter tips
Pointes de pipettes
This range of sterile filter pipette tips feature of the industry’s most efficient aerosol filter assemblies and reliably prevent aerosol transfer.
  • Certified free of DNA, DNase, RNase and endotoxins
  • Tips made from S3 polymer feature a sample saving surface for no hold-ups: Tips retain virtually no fluids, allowing users to dispense truly accurate sample volumes
  • ESP™ reloading system allows reloading 10 trays in as little as 90 seconds: Saves time, lab space and the environment
Ideal for use when performing even the most sensitive assays.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité