VWR®, Stem Thermometer, Calibrated, Electronic, Stainless Steel, 127 mm Probe
Fournisseur: VWR Collection
620-2133EA 55.8 EUR InStock
620-2133
VWR®, Stem Thermometer, Calibrated, Electronic, Stainless Steel, 127 mm Probe
Thermomètres Thermomètres numériques
Digital stem thermometer suitable for field studies, wet lab applications, wash-down areas, food testing and tanks.
  • Will read temperatures in liquids, semi-solids and gases
  • Stainless steel probe easily reaches into cuvettes, test tubes, flasks, beakers and more
  • Includes accredited calibration documentation that meets the requirements of ISO 17025
  • Individually serialised
Certifications: Tested and calibrated in exclusive triple-accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448,01), against equipment whose calibration is traceable to NIST.
Information de livraison: Includes multi-language instructions (English, French, German, Italian, Portuguese and Spanish).
Attention: These electronic stem thermometers are not suitable for solids such as frozen items.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité