Multi-use temperature, humidity and shock data logger, 184 G1
Fournisseur: TESTO
0572.1846
620-2285EA 310 EUR InStock
620-2285
Multi-use temperature, humidity and shock data logger, 184 G1
Enregistreurs de données
Multi-use temperature, humidity and shock data logger is designed for temperature, humidity and impact monitoring in the cold chain and transportation sectors. It is suitable for the food and pharmaceutical industries for the transport of sensitive goods.
  • Measurement rate from one minute to 24 hours
  • Unlimited operating time
  • Clear indication of alarms and alerts
  • Easy setting and automatic PDF reporting via connection to PC
Ideal for monitoring the transport of sensitive products such as electronics, art, pharmaceuticals or foods safely and conveniently.
Certifications: ISO 9001:2008, EN 12830 and HACCP certified. CFR ready. Compliant with GxP.
Information de livraison: Supplied with data logger for shock, humidity and temperature testo 184 G1 for transport monitoring, battery and wall bracket.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité