Room climate monitor, RM100
Fournisseur: EBRO
1348-0001
620-2452EA 186 EUR InStock
620-2452
Room climate monitor, RM100
Systèmes de surveillance Systèmes de surveillance de la température et du CO₂
For monitoring the CO₂ concentration, the ambient temperature and the relative humidity.
  • Three different LEDs (Green, yellow, red) show the current Indoor Air Quality
  • Helps to improve room climate and prevent mold formation
  • Large LCD with backlight for easy reading in the dark
  • Acoustic alarm when CO₂ limit is exceeded
  • MUTE button for alarm on/off
  • For tabletop use or wall-mounting
This smart, compact and easy-to-use device was designed to help people taking care of the Indoor Air Quality. The RM 100 can be widely used in office buildings, schools, exhibition halls, shopping malls, meeting rooms, fitness centers, restaurants and other public places where personal comfort and a healthy environment is important.
Information de livraison: Supplied with 5V-adapter and Factory calibration certificate.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité