Wireless temperature and humidity data loggers, Saveris 2
Fournisseur: TESTO
0572.2103
620-2500EA 374 EUR InStock
620-2500
Wireless temperature and humidity data loggers, Saveris 2
Enregistreurs de données
These data loggers are used for monitoring of temperature and humidity in freezers, refrigerators, storerooms and work environments. These Wi-Fi data loggers reliably record data at defined intervals and transmit the measurement values directly by wireless LAN to the testo Cloud.
  • The measurement values stored in the Cloud can be evaluated anywhere and anytime using an Internet capable smartphone, tablet or PC
  • The display provides information on current measurement values, limit value violations and the remaining battery life
  • Batteries have a lifetime of 24 months and can be exchanged by the user
Storage temperature: −40 to +70 °C
Measuring rate: Basic: 15 minutes (fixed); Advanced: 1 minute to 24 hours (flexible)
Certifications: HACCP International, DIN EN 12830 compliant (Saveris 2-T1/Saveris 2-T2/Saveris 2 set)
Information de livraison: Supplied with USB cable, wall bracket, batteries and calibration protocol. Free access to Testo Cloud (basic). Saveris 2 set also includes two ribbon cable temperature probes, two temperature simulation flasks for filling with glycol.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité