tempmate® M2T Multi-Use Temperature Data Logger, L1 2 °C, H1 8 °C
Fournisseur: tempmate
M2T-100
620-2827EA 60 EUR InStock
620-2827
tempmate® M2T Multi-Use Temperature Data Logger, L1 2 °C, H1 8 °C
Enregistreurs de données
Pre-programmed. Intuitive multi-use temperature logger for reliable low-cost cold chain monitoring. For temperature and humidity recording with internal or external temperature and humidity sensor, multifunction display and automatic PDF report generation.
  • Product-related temperature monitoring along the entire cold chain, in warehouses or fridges
  • Cost-effective and reliable
  • Smart design, intuitive operation
  • Waterproof and food compatible
  • Free available software for individual logger configuration
  • Detailed PDF report including graph and table
  • Multifunction display with battery indicator
tempmate.®-M2 loggers are specially designed for temperature and/or humidity monitoring in pharmaceutical cold chain logistics. As preprogrammed device tempmate.®-M2 can be instantly used to control your temperature sensitive consignment. The PDF report created contains detailed information such as data curve, every single measurement as a table and statistical information. With the optional free available tempbase.®2 software, you can ensure individual logger configuration and detailed data analysis or data management is also possible. Moreover, the device does not require labeling per IATA's dangerous goods regulations.
Certifications: EN 12830, CE, RoHS.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité