M-Clarity™ quality level = MQ100
62107-100G
62107-100GEA
79.5
EUR
InStock
62107-100G
Lewatit® MonoPlus TP 214 résine échangeuse d'ions, macroporeux 400-1250 μm, Supelco®
Lewatit® MonoPlus TP 214
Lewatit® MonoPlus TP 214 is a type of chelating ion-exchange resin.
- Moisture: 43-48%
- Loss: 40-60% loss on drying, 110 °C
- Matrix: Styrene-divinylbenzene (macroporous)
- Matrix active group: Thiourea functional group
- Capacity: 1.0 meq/mL by wetted bed volume
Lewatit® MonoPlus TP 214 may be used as a mercury-specific adsorber resin for the removal of mercury from contaminated groundwater samples prior to its determination by cold vapor atomic absorption spectrometry (CV-AAS). It may also be used as an ion-exchange resin for the determination of platinum and palladium in particles emitted from automobile exhaust catalysts prior to their analysis using cathodic adsorptive stripping voltammetry (CASV).
Chelating resin especially suited for the removal of metals in hydrometallurgical processes and for the extraction/recycling of mercury and silver.