Accessoires
Fournisseur: Dino-Lite / IDCP B.V.
MS12C
630-1159EA 34 EUR InStock
630-1159
Accessoires
Supports de statif
Holders and basic stands for Dino-Lite microscopes.
MS12C: The MS12C consists of a base unit with two insert adapters providing three 'hands free' fixed working distance and magnification options. Ideal for collectors, hobbyists, graphic artists a well as at home or in the classroom. The Dino-Lite digital microscopes simply slides into the base or one of the adapters. The clear acrylic base design allows easy accessibility of tools and light to the observed object. Supplied with two variable view range adapters, transparent.
Suitable for: All Dino-Lite models, except models in AD series and models with polariser.
MS23B: This desktop clamp with flexible gooseneck can be clamped to a desktop or any other surface of up to 3 cm thick. The arm has a wide reach of 35 cm and can be bent and twisted in the required position for easy and convenient inspection of all sorts of objects with the Dino-Lite digital microscope.
Suitable for: All Dino-Lite models.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité