VWR® VisiCam® TC 210, Tablet with Camera
Fournisseur: VWR Collection
630-2678EA 1390 EUR InStock
630-2678
VWR® VisiCam® TC 210, Tablet with Camera
Caméras
This Windows® tablet PC with large LCD touch screen and integrated 5,1 MP camera provides an advanced solution for digital microscopy. For use with any trinocular microscope with C-mount adapter.
  • Easily detachable, can be used as a laptop (keyboard not included)
  • Powerful Intel processor for top performance and speed
  • High resolution and vivid colour graphic display
The imaging software allows live display, image capture, video recording, image size, colour enhancement, brightness gain and data export to PDF or Excel formats.
Tablet specifications:
Operating system: Windows 10, 32-bit
CPU: Intel® Atom™ Z3735F, Quad core
CPU speed: 1,44 GHz
Graphic Card: Intel® HD Graphics 3D Accelerator
RAM 2048 GB DDR3L
Connections: Micro USB-B, USB, microphone, micro SD card reader, mini HDMI and headphones
Network: Wireless, Bluetooth 4,0
W×D×H: 257×105×174 mm
Informations de commande: Keyboard for tablet is sold separately.
Information de livraison: Supplied with PROVIEW software, C-mount adapter for 23 mm tube, calibration slide, USB cable, touch pad.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité