WiFi streamer, Dino-Lite WF-10
Fournisseur: Dino-Lite / IDCP B.V.
WF-10
630-2923EA 299 EUR InStock
630-2923
WiFi streamer, Dino-Lite WF-10
Caméras
WF-10 allows compatible Dino-Lite USB microscopes to be used wirelessly in connection with an iOS (iPhone/iPad) or Android app on any tablet, smartphone, or even in the DinoCapture software for Windows computers.
  • Can be used anywhere to create a WiFi signal for streaming the Dino-Lite live images
  • Can stream from one Dino-Lite microscope to multiple devices
  • Built-in battery is replaceable
Ideal for field work or presentations.
Compatible with: AM/AD4xxx (R4), AM4xx5, AM/AD7xxx (R4), MEDL4xx (R4), MEDL7xx (R4) series.
Please note: The free DinoConnect App needs to be downloaded from the App Store or Google Play.
Commander maintenant
COMMANDE
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité