Handheld digital microscopes, Dino-Lite Basic
Fournisseur: Dino-Lite / IDCP B.V.
AM2111
630-2934EA 129 EUR InStock
630-2934
Handheld digital microscopes, Dino-Lite Basic
Microscopes
These portable microscopes are ideal for general purpose applications, where high image resolution and advanced hardware and software features are not strictly necessary.
  • Economic, easy to use
  • Magnification from 10 to 70× and around 200×
  • Video frame rates up to 30 fps
  • High speed USB 2.0 output
AM2111: Excellent choice for hobbyists and kids (use at schools).
AM3113T: The microscope features measurement functions and the Microtouch button to capture pictures directly from the Dino-Lite. The improved image sensor provides a high quality image with very natural colours and it also offers a high refresh rate for better real time images. It is an affordable solution for industrial, scientific and basic professional use.
Informations de commande: More models − UV/infrared illumination, higher magnification − upon request.
Information de livraison: Supplied with software (DinoCapture 2.0 (Windows) DinoXcope (Mac OS)).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité