USB camera, Moticam® S3
Fournisseur: MOTIC
1100600101091
630-3100EA 683 EUR InStock
630-3100
USB camera, Moticam® S3
Caméras
High Grade sCMOS USB Camera Moticam® S3 for microscopy for an ultimate imaging result.
  • Fast frame rates
  • sCMOS sensors back-illuminated sensor technology
  • USB 3.1 interface
  • Motic Images Plus 3.0 software
A good balance – Moticam S3 has a mid-size resolution of 3 MP, the data volume keeps the balance between speed and resolution. A flexible tool for education and routine labs, for biomedical environments and industry. The sCMOS sensor guarantees state-of-the art imaging results for an unbeatable price.
Informations de commande: To be purchased separately – C-mount adapter not included, this needs to be from the same brand of as your microscope, compatible with your microscope model and fitted with the same size of chip as your Moticam.
Information de livraison: Includes CS ring adapter, USB 3.1 cable, Motic 4-dot calibration slide, also Motic Images Plus 3.0 for Windows/OSX/Linux.
Attention: Minimum computer requirements – 2 GHz dual-core, RAM memory 2 GB, video memory min. 512 MB.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité