USB camera, Moticam® S6
Fournisseur: MOTIC
1100600101111
630-3102EA 856 EUR InStock
630-3102
USB camera, Moticam® S6
Caméras
High Grade Moticam® S6 sCMOS USB microscopy camera for ultimate imaging result.
  • Fast frame rates
  • sCMOS sensors back-illuminated sensor technology
  • USB 3.1 interface
  • Motic Images Plus 3.0 software
The 6 MP resolution of Moticam S6 is targeted to large sized monitors and printing purposes. Presentations to a large audience through video projector and reports in hard copy are performed with ease. The price/performance ratio is also remarkable.
Informations de commande: To be purchased separately – C-mount adapter not included, this needs to be from the same brand as your microscope, compatible with your microscope model and fitted with the same size of chip as your Moticam.
Information de livraison: Includes CS ring adapter, USB 3.1 cable, Motic 4-dot calibration slide, and Motic Images Plus 3.0 for Windows/OSX/Linux.
Attention: Minimum computer requirements – 2 GHz dual-core, RAM memory 2 GB, video memory min. 512 MB.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité