USB camera, Moticam® Pro S5 Plus
Fournisseur: MOTIC
1100600101121
630-3103EA 2690 EUR InStock
630-3103
USB camera, Moticam® Pro S5 Plus
Caméras
High Grade sCMOS USB Camera Moticam® Pro S5 Plus for microscopy for an ultimate imaging result for fluorescence applications.
  • Fast frame rates
  • sCMOS sensors back-illuminated sensor technology
  • USB 3.1 interface
  • Ideal for fluorescence applications
  • Motic Images Plus 3.0 software
An Ultimate Solution, Still Affordable – Moticam Pro S5 Plus carries a large 2/3” sensor with 5MP resolution at a maximized frame rate of 70 fps* in full resolution. This camera covers all important aspects of imaging: resolution for detail information, speed for fast phenomena and a large sensor to cover extended areas in a single shot.
Informations de commande: To be purchased separately – C-mount adapter not included to be purchased from the same brand of your microscope, compatible with your microscope model and fitting with the size of the chip of your Moticam.
Information de livraison: Includes CS ring adaptor, USB 3.1 cable, Motic 4-dot calibration slide, Motic Images Plus 3.0 for Windows/OSX/Linux.
Attention: Minimum computer requirements – 2 GHz dual-core, RAM memory 2 GB, video memory min. 512 MB.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité