Upright laboratory microscopes, DM2000 LED
Fournisseur: LEICA MICROSYSTEMS
11888843_P1 11888843_P2 11888843_P3 11888843_P4
630-3182EA 6480 EUR InStock
630-3182 630-3183 630-3184 630-3185
Upright laboratory microscopes, DM2000 LED
Microscopes
With their sophisticated modular design and high performance fluorescence, Leica DM2000 LED microscopes are ideal for complex tasks in pathology, cytology and many other applications. For special diagnostic requirements, the microscope is certified for in-vitro diagnostic (IVD) like in-vitro fertilisation (IVF).
  • Height adjustable focus knobs provide the optimal choice of hand position for a relaxed hand position
  • Adjustable eye tubes promotes correct posture
  • Symmetrical operation from either left or right to facilitate alternating users
  • Telescopic coaxial
More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité