Hybrid microscope (upright + inverted), ECHO Rebel
Fournisseur: DISCOVER ECHO
RBLTEW31
630-3346EA 7285 EUR InStock
630-3346
Hybrid microscope (upright + inverted), ECHO Rebel
Microscopes
Rebel is designed around the user to challenge the status quo. Designed from the ground up to improve microscopy.
  • ECHO Pro: powerful yet simple software reimagined to collaboratively view, capture and share images
  • Ultra-high resolution 13" touch display
  • 12 MP CMOS colour camera
  • Touch interface is very easy to use
  • Plan achromat and apochromat options available
  • XY mechanical stage option available
  • Ergonomic biosafety cabinet and adjustable height table options available
Versatility: Hybrid microscope easily converts between upright and inverted configurations. Brightfield and phase contrast modes available. Eyepiece option available as well.
Productivity: Our award-winning touch interface is incredibly easy to use. Eyepieces have been replaced with a 13" iPad with retina display to view and capture stunning crystal-clear images.
Rebel True Ergo Table and Rebel Logic + Biosafety cabinet by Labconco bring the microscope stage down flush to the tabletop, setting a new precedent in microscope ergonomics.
Informations de commande: Microscope includes a 2 year warranty on electrical, and 5 year warranty on mechanical parts and optics. Contact your local Avantor representative for available options or to demo the latest innovations in microscopy.
Information de livraison: Includes microscope body, iPad with locking stand, 12 MP CMOS colour camera, power cord, microscope cover, and lens cloth.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
SERVICES
Services: Contact your VWR representative for product demonstrations.
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité