Imager model organism package, THUNDER
Fournisseur: LEICA MICROSYSTEMS
10450921_P1
630-3394EA 0 EUR InStock
630-3394
Imager model organism package, THUNDER
Microscopes
The THUNDER imager model organism allows fast and easy 3-D exploration of whole organisms (e.g., Drosophila, C. elegans, zebrafish, plants and mice) for developmental or molecular biology research. Eliminate the hassle of out-of-focus blur while maintaining the capabilities and ease of use typical for Leica stereo microscopes.
Takes research to the next level with improved image quality to get the most information from your precious model organisms with THUNDER imager model organism.
Certifications: CE and CAS.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité