Guides de lumière: Cols de cygne
Fournisseur: SCHOTT AG Lighting and imaging
154101 154202 154302 170101 170202 120402 170402
631-0021EA 197 EUR InStock
631-0021 631-0022 631-0023 631-0001 631-0002 631-1024 SCOC170402
Guides de lumière: Cols de cygne
Éclairages Guides de lumière
Fibre optic illumination for stereo microscopy. To be used with cold light sources.
In all its microscopy light guides, SCHOTT uses the new environmentally friendly glass optical fibers PURAVIS®. Not just the fibers themselves, but also the entire proprietary manufacturing process are lead free and avoid the use of arsenic and antimony. Due to superior physical and chemical properties, PURAVIS® glass optical fibers provide a high performance, superior longevity, improved transmission in the near UV and outstanding transmission of white light.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité