Cryostat, model 6250
Fournisseur: DAKEWE MEDICAL
130.012.008-00
631-1288EA 18610 EUR InStock
631-1288
Cryostat, model 6250
Microtomes et groupes de microtomes Microtomes
Cryostat features a one-time injected model cover, anti-fog heating glass design, and a chromatic liquid crystal touch screen.
  • Modular control system
  • Can be controlled by touch screen or joystick
  • Defrost automatically/manually
  • Hibernation automatically/manually, wake up in 15 minutes
  • UV sterilisation
  • Semi conductor refrigerant
  • Section number and thickness calculation
Sectioning Quality: It has light hand wheel with locking device. Specimen clamp enables to adjust triaxially with a center block. High percision guide rails makes transmission smooth, durable and be able to cut hard sample. Retraction function is available.
Freezing chamber: Extra large freezing chamber with wide freezing shelf may hold 20 specimen discs meanwhile. It also has waste tray and instrument rack.
Blade holder: It has wide and narrow blade options. Adjustable eccentric shaft ensures blade clamp tight. Blade clamp can be moved horizontically on the guide rail for safety and making use of the blade. Anti-rolling plate is horizontically and verticaly adjustable and also able to adjust angle and gap with pressing plate. A pushing rod pops the blade out easily.
Informations de commande: Contact your Avantor representative for more information.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité