NMR valve with tube attached
Fournisseur: GLASS PRECISION ENGINEERING
S-5-400-JY-7
634-1304EA 197 EUR InStock
634-1304
NMR valve with tube attached
Tubes Tubes RMN
Norell select series NMR tube with 5 mm NMR valve.
  • Negative pressure/high vacuum, leak tested to 10⁻⁹ mbar L/sec
  • Internal positive pressure 5 bar (72 psi)
Handle your NMR sample without flame sealing your tubes. Teflon covered O-Ring eliminates material incompatibilities and completely greaseless Teflon assembly means the tube is easy to use and to disassemble for cleaning. Includes female joint for quick attachment to your vacuum rack.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité