Coaxial inserts
Fournisseur: GLASS PRECISION ENGINEERING
NI5CCI-B
634-1309EA 123 EUR InStock
634-1309
Coaxial inserts
Inserts
Made from low expansion borosilicate glass (Pyrex® or equivalent ASTM Type 1 Class A glass).
A coaxial insert consists of an upper glass alignment tube with a precise outer diameter that fits very closely within the matching 5 or 10 mm thin wall outer glass NMR tube, ensuring accurate coaxiality and centering of the insert within the outer NMR tube. The lower glass stem tube, of a smaller diameter, typically holds an external lock or reference material. The smaller diameter stem tube is precision sealed to the upper alignment tube using a highly accurate, state-of-the-art glass working lathe, maintaining the coaxial alignment necessary for the acquisition of high quality spectra. The accompanying table presents the most commonly used sizes for Bruker and Varian probes.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité